Learn More
Enpiroline, TRC
Product Code. 30383955
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383955 100 mg
30370340 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30383955 Supplier Toronto Research Chemicals Supplier No. TRCE557650100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 66364-73-6
Molecular Formula C19 H18 F6 N2 O
Molecular Weight (g/mol) 404.35
Synonym αr)-Rel-α-(2R)-2-Piperidinyl-2-(Trifluoromethyl)-6-[4-(Trifluoromethyl)Phenyl]-4-Pyridinemethanol, (R*,R*)-(±)-α-2-Piperidinyl-2-(Trifluoromethyl)-6-[4-(Trifluoromethyl)Phenyl]-4-Pyridinemethanol
SMILES O[C@@H]([C@H]1Ccccn1)C2Cc(Nc(C2)C(F)(F)F)C3Ccc(Cc3)C(F)(F)F
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C19H18F6N2O/c20-18(21,22)13-6-4-11(5-7-13)15-9-12(10-16(27-15)19(23,24)25)17(28)14-3-1-2-8-26-14/h4-7,9-10,14,17,26,28H,1-3,8H2
Formula Weight 404.1323
Chemical Name or Material Enpiroline