missing translation for 'onlineSavingsMsg'
Learn More

Enilconazole, Mikromol™

Product Code. 18081270
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18081270 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18081270 Supplier Mikromol Supplier No. MM1228.00
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Enilconazole
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Imazalil
CAS 35554-44-0
Analyte or Component Names Imazalil
Shipping Condition Room Temperature
Molecular Formula C14 H14 Cl2 N2 O
Certifications/Compliance ISO 17025
Synonym Enilconazole, 1-[(2RS)-2-(2,4-Dichlorophenyl)-2-(prop-2-enyloxy)ethyl]-1H-imidazole, Imazalil, 1H-Imidazole, 1-[2-(2,4-dichlorophenyl)-2-(2-propen-1-yloxy)ethyl]-, 1H-Imidazole, 1-[2-(2,4-dichlorophenyl)-2-(2-propenyloxy)ethyl]- (9CI), 1-[2-(2,4-Dichlorophenyl)-2-(2-propen-1-yloxy)ethyl]-1H-imidazole, (R,S)-1-[2-(2,4-Dichlorophenyl)-2-(2-propenyloxy)ethyl]-1H-imidazole, (RS)-1-[2-(Allyloxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole, (+/-)-Imazalil, 1-[2-(2,4-Dichlorophenyl)-2-(2-propenyloxy)ethyl]-1H-imidazole, Amolden MP 100, Bromazil, CGA 41333, Clinafarm, Deccosil, Deccozil, Deccozil S 75, FloPro IMZ, Florasan, Freshgard, Fungaflor, Fungazil, Imaverol, Magnate, NuZone, R 23979
InChI Formula InChI=1S/C14H14Cl2N2O/c1-2-7-19-14(9-18-6-5-17-10-18)12-4-3-11(15)8-13(12)16/h2-6,8,10,14H,1,7,9H2
SMILES Clc1ccc(C(Cn2ccnc2)OCC=C)c(Cl)c1
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-prop-2-enoxyethyl]imidazole
Molecular Weight (g/mol) 297.18
Quantity 250 mg
Formula Weight 296.0483 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.