Learn More
Lonza™ Endotoxin
Product Code. 11600461
Change view
Click to view available options
Quantity:
5 viales
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11600461 5 viales Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11600461 Supplier Lonza™ Supplier No. 193783
  • ONLINE EXCLUSIVE PRICE
    903.00 EUR
    800.07 EUR / Pack of 1
    Save 102.93 EUR
    11% Off
 Restricted Item
Estimated Shipment: 23-11-2026
High potency endotoxin intended for use in endotoxin removal system challenges, i.e., depyrogenation ovens, and other spiking studies.
  • High potency
  • Does not contain filler
  • Can be used for oven validation studies
TRUSTED_SUSTAINABILITY
Specifications
Concentration 1.25 million EU/vial
Endotoxin Level 1.25 million EU/vial
For Use With (Application) Endotoxin removal
Quantity 5 viales