missing translation for 'onlineSavingsMsg'
Learn More

Enclomiphene citrate, MedChemExpress

Product Code. p-200099240
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16457647 5 mg 5mg
16431147 50 mg 50mg
16415676 100 mg 100mg
16404898 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16457647 Supplier Medchem Express Supplier No. HY118861A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Enclomiphene ((E)-Clomiphene) citrate is a potent and orally active non-steroidal estrogen receptor antagonist, with antioestrogenic property. Enclomiphene citrate can be used for the research of ovarian dysfunction, testosterone deficiency, male hypogonadism and type 2 diabetes.

Enclomiphene ((E)-Clomiphene) citrate is a potent and orally active non-steroidal estrogen receptor antagonist, with antioestrogenic property. Enclomiphene citrate can be used for the research of ovarian dysfunction, testosterone deficiency, male hypogonadism and type 2 diabetes.

Specifications

Chemical Name or Material Enclomiphene citrate
CAS 7599-79-3
Color White
Purity Grade Notes Research
Molecular Formula C32H36ClNO8
Quantity 5 mg
Synonym (E)-Clomiphene citratetrans-Clomiphene citrateEnclomifene citrate
Solubility Information DMSO : ≥ 50 mg/mL (83.60 mM)
SMILES Cl/C(C1=CC=CC=C1)=C(C2=CC=C(OCCN(CC)CC)C=C2)\C3=CC=CC=C3.O=C(CC(C(O)=O)(O)CC(O)=O)O.[(E)]
Molecular Weight (g/mol) 598.08
Formula Weight 598.08
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.