missing translation for 'onlineSavingsMsg'
Learn More

Enalaprilat dihydrate, MedChemExpress

Product Code. 16474307
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16474307

Brand: Medchem Express HYB0231

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Enalaprilat dihydrate (MK-422), the active metabolite of the oral prodrug Enalapril, is a potent, competitive and long-acting angiotensin-converting enzyme (ACE) inhibitor, with an IC50 of 1.94 nM. Enalaprilat dihydrate can be used for the research of hypertension.

Enalaprilat dihydrate (MK-422), the active metabolite of the oral prodrug Enalapril, is a potent, competitive and long-acting angiotensin-converting enzyme (ACE) inhibitor, with an IC50 of 1.94 nM. Enalaprilat dihydrate can be used for the research of hypertension.

Specifications

Chemical Name or Material Enalaprilat dihydrate
CAS 84680-54-6
Color White
Purity Grade Notes Research
Molecular Formula C18H28N2O7
Quantity 50 mg
Synonym MK-422
Solubility Information DMSO : 100 mg/mL (260.13 mM; Need ultrasonic) ∣H2O : 12.5 mg/mL (32.52 mM; Need ultrasonic)
SMILES O=C(O)[C@H]1N(C([C@H](C)N[C@H](C(O)=O)CCC2=CC=CC=C2)=O)CCC1.O.O
Molecular Weight (g/mol) 384.42
Formula Weight 384.42
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.