Learn More
Merck MultiScreen™ 96-Well Assay Plates for Sample Preparation
Product Code. 10607891 Shop All Merck Products
Change view
Click to view available options
Pore Size:
0.22 μm
0.45 μm
0.65 μm
1.2 μm
Color:
Clear
Opaque
Material:
ABS Plastic
Barex
Styrene
Quantity:
10/Pk.
50/Pk.
Unit Size:
Pack of 10
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Color Material Quantity unitSize
10607891 0.22 μm Clear ABS Plastic 10/Pk. Pack of 10
10470211 1.2 μm Clear Styrene 50/Pk. Pack of 50
10370031 0.65 μm Clear Styrene 50/Pk. Pack of 50
10711801 0.45 μm Clear Styrene 10/Pk. Pack of 10
10054540 0.45 μm Opaque Barex 50/Pk. Pack of 50
10658271 0.45 μm Clear Styrene 50/Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10607891 Supplier Merck Supplier No. MAGVS2210
  • ONLINE EXCLUSIVE PRICE
    488.00 EUR
    454.00 EUR / Pack of 10
    Save 34.00 EUR
    6% Off
View all promotions
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
For the isolation, purification, and preparation of samples prior to testing in drug discovery processes and protein and DNA applications
Formats are suitable for removal of large particulates, resin-based separations, solvent applications, and protein/DNA concentration or purification. Versatile separation system combines a microplate with Millipore's membrane filtration to simplify and optimize drug discovery techniques, biochemical assays, and traditional sample preparation.
  • Use with a vacuum manifold, automated liquid-handling system, or microplate scintillation counter—entire assays can be performed on one plate
  • Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Patented underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days
  • Also ensures quantitative filtrate transfer to a receiver plate placed in bottom of manifold
Choice of filter materials
  • For high protein binding applications, use plates with Immobilon™-P PVDF or mixed cellulose esters For high nucleic acid binding, use plates with mixed cellulose esters, glass fiber, or positively charged DEAE membranes For charged biomolecule separations, choose from positively charged DEAE or negatively charged phosphocellulose membranes
  • For low biomolecule binding, use plates with hydrophilic Durapore™ PVDF, Biopore™ PTFE, or polycarbonate membranes
  • For assays that require solvent resistance, use plates with hydrophilic PTFE membranes
Each microporous membrane interacts distinctly with different proteins or biomolecules; therefore, it may be necessary to test different membrane types for each assay.
†MultiScreenHTS version features a recessed underdrain and rigid sidewalls.
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Membrane Hydrophilic PVDF
Array 8 x 12
Pore Size 0.22 μm
Color Clear
Product Type MultiScreen-GV Plate
Material ABS Plastic
No. of Wells 96
Sterility Sterile
Filtration Area 0.28cm2
For Use With (Application) Sample prep or suspension cell culture, YAC culture
Quantity 10/Pk.