Description
The EMA 502 Elemental Analyzer is the accurate and reliable solution for the determination of carbon, hydrogen, nitrogen, sulfur, and oxygen in various industrial sectors such as pharma and life science, organic chemistry, petrochemistry and energy, environmental, agronomy, food and feed. The EMA 502 offers consistent performance, versatility, ease-of-use and premium features for combustion analysis following international standards: AOAC, AACC, ASBC, ISO, DIN, IFFO, OIV, ASTM, EPA.
ONE SOLUTION FOR MULTIPLE ANALYTICAL CHALLENGES
- 30-position electronic autosampler expandable with extra 3 discs to ensure maximum productivity
- Seamless switch from CHNS to O mode with exclusive connection panel, without the need for external modules
- Helium and Argon as carrier gas
VERSATILITY AND PRODUCTIVITY
- The exclusive TCD of the EMA 502 ensures maximum accuracy and reliability
- Precise determination from a few ppm to 100% for solid, paste and liquid samples
- The Gas Chromatographic technology ensures the complete separation of all elements and real-time peak data
SUPERIOR RELIABILITY AND ACCURACY
- Intuitive operation with the powerful EMASoft™ software
- Monitor the analytical process with the real-time graph
- Benefit from a comprehensive reporting system
UNMATCHED EASE-OF-USE
- The exclusive connectivity to VELP Ermes Cloud Platform projects your lab to a data-rich environment with premium remote service support resulting in the highest system uptime.
Specifications
Specifications
| Type | Elemental Analyzer |
| Detector Type | Thermal Conductivity Detector |
| For Use With (Application) | Organic Chemistry, Environmental Science, Petro-Chemistry, Material Characterization, Pharmaceuticals |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.