missing translation for 'onlineSavingsMsg'
Learn More

Elvitegravir, TRC

Product Code. 30009420
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009420 10 mg 10mg
30005000 25 mg 25mg
30009132 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009420 Supplier Toronto Research Chemicals Supplier No. TRCE50900010MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 697761-98-1
Molecular Formula C23 H23 Cl F N O5
Molecular Weight (g/mol) 447.88
Synonym Elvitegravir, 3-Quinolinecarboxylic acid, 6-[(3-chloro-2-fluorophenyl)methyl]-1,4-dihydro-1-[(1S)-1-(hydroxymethyl)-2-methylpropyl]-7-methoxy-4-oxo-, 6-[(3-Chloro-2-fluorophenyl)methyl]-1,4-dihydro-1-[(1S)-1-(hydroxymethyl)-2-methylpropyl]-7-methoxy-4-oxo-3-quinolinecarboxylic acid, 1,4-Dihydro-6-(3-chloro-2-fluorobenzyl)-7-methoxy-1-[(1S)-1-(hydroxymethyl)-2-methylpropyl]-4-oxo-3-quinolinecarboxylic acid, 6-(3-Chloro-2-fluorobenzyl)-1-[(S)-1-hydroxymethyl-2-methylpropyl]-7-methoxy-4-oxo-1,4-dihydroquinoline-3-carboxylic acid, GS 9137, JTK 303, Vitekta
IUPAC Name 6-[(3-chloro-2-fluorophenyl)methyl]-1-[(2S)-1-hydroxy-3-methylbutan-2-yl]-7-methoxy-4-oxoquinoline-3-carboxylic acid
SMILES COc1cc2N(C=C(C(=O)O)C(=O)c2cc1Cc3cccc(Cl)c3F)[C@H](CO)C(C)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C23H23ClFNO5/c1-12(2)19(11-27)26-10-16(23(29)30)22(28)15-8-14(20(31-3)9-18(15)26)7-13-5-4-6-17(24)21(13)25/h4-6,8-10,12,19,27H,7,11H2,1-3H3,(H,29,30)/t19-/m1/s1
Formula Weight 447.1249
Percent Purity >95%
Chemical Name or Material Elvitegravir
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.