missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies ELISA Diluent

Product Code. 17100002
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 120 mL
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17100002 2 x 120 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17100002 Supplier STEMCELL Technologies Supplier No. 01916
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Diluent for ELISA samples, standards, and detection antibodies

  • ELISA Diluent is intended for dilution of samples, standards, and detection antibodies in ELISA assays
  • By preventing the cross-linking of heterophilic antibodies, ELISA Diluent minimizes interference and false-positive signals in ELISA reactions
  • ELISA Diluent is required for use with the Human IgE ELISA Antibody Pair Kit (Catalog #01993) when serum or plasma samples are used
  • NOTE: Do not use ELISA Diluent for dilution of streptavidin conjugates

Specifications

CAS 26172-55-4
Description Cell Type: B Cells, Hybridomas
For Use With (Application) Hybridoma Generation; Immunology
Quantity 2 x 120 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.