Learn More
Bibby Scientific™ Electrothermal™ Replacement Spare Heater Element
Product Code. 11758422
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11758422 100 mL Each
11768422 250 mL Each
11778422 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11758422 Supplier Bibby Scientific™ Supplier No. 1008642
Only null left
Specifications
Capacity (Metric) 100 mL
For Use With EMEA3/6/0100
Voltage 230 V
Wattage 60w
Product Type Replacement Spare Heater Element
Frequency 50 Hz