Learn More
Thermo Scientific™ Electroporation Cuvettes
Product Code. 10227570 Shop All MBP Molecular BioProducts Products
Change view
Click to view available options
Width (Metric) Gap:
1 mm
2 mm
4 mm
Unit Size:
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Gap Unit Size
10227570 2 mm Pack of 50
10339270 1 mm Pack of 50
12338212 1 mm Pack of 50
12348212 4 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10227570 Supplier Thermo Scientific™ Supplier No. 5520
  • ONLINE EXCLUSIVE PRICE
    256.00 EUR
    235.60 EUR / Pack of 50
    Save 20.40 EUR
    7% Off
View all promotions
Estimated Shipment: 21-12-2026
Increase the ease of sample identification and labelling with these premium-grade polycarbonate electroporation cuvettes, which have frosted, colored caps.
Increase the ease of sample identification and labeling with premium-grade polycarbonate Thermo Scientific™ Electroporation Cuvettes, which have frosted, colored caps.
  • Available in 1mm, 2mm, or 4mm gap sizes
  • Pre-sterilized and individually wrapped
  • Compatible with all common electroporators
TRUSTED_SUSTAINABILITY
Specifications
Sterility Sterile
Type Electroporation Cuvette
Width (Metric) Gap 2 mm