Learn More
Thermo Scientific Electronic Handheld Crimper for 20mm Flip-off Crimp Caps
Brand:  Thermo Scientific 60180-ECR20FO
1004.31 EUR valid until 2026-08-31
BEST PRICE on Promo! Use promo code "28036" to get your promotional price.
Electronic Handheld Crimper for 20mm Flip-off Crimp Caps
Product Code. 16450955
  • 1057.17 EUR / Pack of 1
Spotlight Offers
Use promo code 28036
View all promotions
Estimated Shipment: 11-09-2026
Certificates
Promotions
Promotions
Promo
Expires: 2026-08-31
Spotlight Offers
Promo Code: 28036
Discover Spotlight promotions, featuring exclusive offers on laboratory chemicals, equipment, consumables, and essential lab supplies for a limited time.
Applying Promo and Adding...