Learn More
Sherwood Scientific™ Electrode Polish Vial
Product Code. 12376307
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12376307 500 mL 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12376307 Supplier Sherwood Scientific™ Supplier No. 00156205
  • ONLINE EXCLUSIVE PRICE
    39.80 EUR
    30.16 EUR / 25g
    Save 9.64 EUR
    24% Off
 Restricted Item
Estimated Shipment: 10-12-2026