missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elbasvir, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Elbasvir |
| CAS | 1370468-36-2 |
| Quantity | 100 mg |
| Shipping Condition | No special requirements |
| Product Line | Life science |
| Molecular Formula | C49 H55 N9 O7 |
| Physical Form | Powder |
| Color | White to Light-yellow |
| Purity | ≥94% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction