missing translation for 'onlineSavingsMsg'
Learn More

EDTA Calcium Disodium Salt Hydrate, TRC

Product Code. 30138509
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138509 50 g 50g
30134546 10 g 10g
30010428 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138509 Supplier Toronto Research Chemicals Supplier No. TRCE33510850G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-33-9
Molecular Formula C10 H12 Ca N2 O8 . 2 Na
Molecular Weight (g/mol) 374.27
Synonym Sodium calcium edetate, Disodium [(ethylenedinitrilo)tetraacetato]calciate(2-)
SMILES [Na+].[Na+].O=C1CN23CCN45CC(=O)[O-][Ca+2]24([O-]C(=O)C5)([O-]C(=O)C3)[O-]1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C10H16N2O8.Ca.2Na/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;/q;+2;2*+1/p-4
Formula Weight 374.0015
Chemical Name or Material Sodium calcium edetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.