missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDR1/PHC1 Antibody (1F3F3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
203.00 EUR - 552.00 EUR
Specifications
| Antigen | EDR1/PHC1 |
|---|---|
| Clone | 1F3F3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18600786
|
Novus Biologicals
NBP2-37371-0.025ml |
0.025 mL |
203.00 EUR
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18136108
|
Novus Biologicals
NBP2-37371 |
0.1 mL |
552.00 EUR
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EDR1/PHC1 Monoclonal specifically detects EDR1/PHC1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| EDR1/PHC1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| Unconjugated | |
| RUO | |
| Human | |
| early development regulator 1 (homolog of polyhomeotic 1), Early development regulatory protein 1, EDR1, hPH1, PH1, polyhomeotic homolog 1 (Drosophila), polyhomeotic-like 1, polyhomeotic-like 1 (Drosophila), polyhomeotic-like protein 1, RAE28 | |
| PHC1 | |
| IgG1 | |
| Affinity Purified |
| 1F3F3 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS with 0.05% Sodium Azide | |
| 1911 | |
| Purified recombinant fragment of human PHC1 (AA: 758-1004) expressed in E. Coli. | |
| Primary | |
| 105.5 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title