missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDR1/PHC1 Antibody (1F3F3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37371
This item is not returnable.
View return policy
Description
EDR1/PHC1 Monoclonal specifically detects EDR1/PHC1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| EDR1/PHC1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| early development regulator 1 (homolog of polyhomeotic 1), Early development regulatory protein 1, EDR1, hPH1, PH1, polyhomeotic homolog 1 (Drosophila), polyhomeotic-like 1, polyhomeotic-like 1 (Drosophila), polyhomeotic-like protein 1, RAE28 | |
| Mouse | |
| 105.5 kDa | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 1911 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| 1F3F3 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| PHC1 | |
| Purified recombinant fragment of human PHC1 (AA: 758-1004) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction