Description
- Advanced protection: EDGE™ 48-707 cut and abrasion-resistant gloves provide ANSI cut level A6 and EN 388 ISO cut level F, which afford a very high level of cut protection
- Heightened grip and dexterity: Owing to their palm PU coating, these protective gloves combine good grip as well as dexterity, and are suitable for dry or slightly oily environments
- Reduced risks: In addition, EDGE™ 48-707 gloves meet electrostatic discharge protection needs and feature anti-static capabilities.
- Added features: These grey hand protection gloves also possess a seamless liner and knitted wrist, which deliver a secure fit for use in applications that require high cut resistance
Specifications
Specifications
| Coating Material | Polyurethane |
| Color | Black |
| Cut Resistance Rating | Level F |
| Hand | Specific |
| Latex Free | Yes |
| Material | Polyester, Fiberglass, Spandex, Nylon, HPPE |
| Product Type | Industrial Gloves |
| Size | 6 |