missing translation for 'onlineSavingsMsg'
Learn More

Edetate Calcium Disodium Dihydrate, TRC

Product Code. 30136235
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136235 50 g 50g
30132492 10 g 10g
30006092 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136235 Supplier Toronto Research Chemicals Supplier No. TRCE55559350G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6766-87-6
Molecular Formula C10H12CaN2O8 . 2H2O . 2Na
Molecular Weight (g/mol) 410.3
Synonym Edetate Calcium Disodium(Secondary Standards Traceble To Usp)
SMILES O.O.[Na+].[Na+].[Ca+2].[O-]C(=O)CN(CCN(CC(=O)[O-])CC(=O)[O-])CC(=O)[O-]

Specifications

Quantity 50 g
InChI Formula InChI=1S/C10H16N2O8.Ca.2Na.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;;2*1H2/q;+2;2*+1;;/p-4
Formula Weight 410.0226
Chemical Name or Material Edetate Calcium Disodium Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.