Learn More
BRAND™ Economy Discharge Tip For Compact Burettes
Product Code. 10054240 Shop All Brand Products
Change view
Click to view available options
Material:
Polypropylene
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10054240 Polypropylene Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10054240 Supplier BRAND™ Supplier No. 11600
  • ONLINE EXCLUSIVE PRICE
    2.50 EUR
    2.10 EUR / Pack of 1
    Save 0.40 EUR
    16% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Includes screw cap
  • For PTFE stopcocks
  • For SILBERBRAND, AR-GLAS™ Burette tubes
  • TRUSTED_SUSTAINABILITY
    Specifications
    For Use With (Equipment) PTFE Stopcock
    Material Polypropylene
    No. per Pack 1
    Stopcock Material PTFE
    Type Discharge Tip