Learn More
Fisherbrand™ Mechanical Benchtop Counter
Product Code. 11334095 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Description:
6-key Counter
9-key Counter
Length (Metric):
250 mm
320 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Length (Metric) Unit Size
11334095 6-key Counter 250 mm Each
11344095 9-key Counter 320 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11334095
  • ONLINE EXCLUSIVE PRICE
    247.00 EUR
    198.14 EUR / Pack of 1
    Save 48.86 EUR
    19% Off
View all promotions
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
Depress the keys in the appropriate blood cell category to keep track of counts. Mechanical Benchtop Counter is an economical solution for counting blood cells, bacterial colonies and other types of repetitive counting.
  • Each key records up to 999
  • Total window keeps track of total number of strokes made on all other keys
  • Bell rings when total reaches 100 blood cell key labels
    • HS5360A:
      • Baso., Eosin., Mono., Lymph., Poly., and Total,
      • Measures 250 x 76 x 56 mm
    • HS5360B:
      • Baso, Eosin., Myelo., Juven., Stab., Seg., Lymph., Mono., and Total,
      • Measures 320 x 76 x 56 mm
Specifications
Product Type Mechanical Counter
Display Type Mechanical
Description 6-key Counter
Model HS5360A
Quantity 1 Ea.
Height (English) 2.2 in.
Height (Metric) 56 mm
Length (English) 9.8 in.
Length (Metric) 250 mm
Width (English) 3 in.
Width (Metric) 76 mm
Includes Labels: Baso., Eosin., Mono., Lymph., Poly., and Total