Learn More
Sharpsguard™ Eco Waste Container with Needle Remover
Product Code. 17714844
Change view
Click to view available options
Color:
Orange
Yellow
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17714844 Orange Each
17724844 Yellow Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Product Code. 17714844 Supplier Sharpsguard™ Supplier No. DD442NROLR
  • ONLINE EXCLUSIVE PRICE
    3.00 EUR
    2.46 EUR / Pack of 1
    Save 0.54 EUR
    18% Off
Estimated Shipment: 22-12-2026
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
The new Sharpsguard™ eco containers, made using at least 40% recycled plastic that is used for the disposal of sharps that have not been contaminated with medicines.
  • Sharpsguard™ eco 0.5 with needle remover, is ideal for community healthcare workers and easily fitted into a bag or glove compartment.
  • This container is already supplied with the lid assembled. Details about how to lock the container and keep the users and contents safe can be found on the label.
  • Whilst in use containers should be positioned safely using the base plate 0.5 from our POUDS™ range.
  • Apart from a difference in shade of color and an added 'R' on the end of the product code, there is no difference to the safe and easy to use range that you know and trust.
  • This container is for disposal by incineration or other authorized process and is designed for the consignment of waste categorized by EWC codes: 18-01-01, 18-01-03, 18-02-01, 18-02-02.
Specifications
Capacity (Metric) 0.4 L
Color Orange
Dimensions (L x W x H) 79 x 45 x 206 mm
Opening Size (Metric) 26 x 30 mm
Weight (Metric) 88 g
Includes Needle Remover