Learn More
Sharpsguard™ Eco 1 Waste Container
Product Code. 17784854
Change view
Click to view available options
Color:
Orange
Yellow
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17784854 Yellow Each
17774854 Orange Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17784854 Supplier Sharpsguard™ Supplier No. DD477YLR
  • ONLINE EXCLUSIVE PRICE
    2.00 EUR
    1.72 EUR / Pack of 1
    Save 0.28 EUR
    14% Off
Estimated Shipment: 02-11-2026
The new Sharpsguard™ eco 1 container, made using at least 40% recycled plastic that is used for the disposal of sharps that have been contaminated with medicines, but not cytotoxic.
  • Sharpsguard™ eco 1 is ideal for phlebotomists. Its push-down, one-piece lid incorporates a vacutainer remover, and will fit securely into our POUDS™ tray range and other carrying trays.
  • This container is already supplied with the lid assembled. Details about how to lock the container and keep the users and contents safe can be found on the label.
  • This container is for disposal by incineration or other authorized process and is designed for the consignment of waste categorized by EWC codes: 18-01-01, 18-01-03, 18-02-01, 18-02-02.
Specifications
Capacity (English) 0.84 L
Color Yellow
Dimensions (L x W x H) 204 x 68 x 119 mm
Opening Size (Metric) 70 x 32 mm
Quantity 1 Ea.
Width (Metric) 153 g