Learn More
Ecgonidine Ethyl Ester, TRC
Product Code. 30135497
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135497 250 mg 250mg
30004553 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30135497 Supplier Toronto Research Chemicals Supplier No. TRCA795290250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 73045-45-1
Molecular Formula C11H17NO2
Molecular Weight (g/mol) 195.26
Synonym (-)-Anhydroecgonine Ethyl Ester, (R)-Anhydroecgonine Ethyl Ester, Anhydroecgonine Ethyl Ester, 8-Azabicyclo[3.2.1]oct-2-ene-2-carboxylic Acid 8-Methyl-Ethyl Ester, Ecgonidine Ethyl Ester, Ethyl Ecgonidine, Methylecgonidine
IUPAC Name ethyl (1R,5S)-8-methyl-8-azabicyclo[3.2.1]oct-2-ene-2-carboxylate
SMILES CCOC(=O)C1=CC[C@@H]2CC[C@H]1N2C
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C11H17NO2/c1-3-14-11(13)9-6-4-8-5-7-10(9)12(8)2/h6,8,10H,3-5,7H2,1-2H3/t8-,10-/m1/s1
Formula Weight 195.1259
Chemical Name or Material Ecgonidine Ethyl Ester