Learn More
Invitrogen™ eBioscience™ Mouse Intracellular (cytoplasmic/cytokine) Protein Flow Cytometry Workflow Kit
Product Code. 17717473
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17717473 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17717473 Supplier Invitrogen™ Supplier No. A53015
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The eBioscience™ Mouse Intracellular (cytoplasmic/cytokine) Protein Flow Cytometry Workflow Kit contains all of the normal buffers, blockers, and beads needed to complete a standard flow cytometry experiment for staining intracellular cytoplasmic and cytokine targets in mouse cell suspensions.
  • Kit contains all of the normal buffers, blockers and beads to complete a standard flow cytometry experiment for staining intracellular cytoplasmic and cytokine targets in mouse cell suspensions
  • When combined with experiment-specific antibodies, the reagents within this kit have been optimized to work together to detect mouse intracellular cytoplasmic and cytokine targets by flow cytometry
  • Effective compensation controls
  • Fc receptor blocking
  • Non-antibody, non-protein based blocking solution for reducing background staining from fluorophores
  • Antibody binding buffer minimizes non-specific binding of antibodies
  • Fixation buffer for storage of stained samples
  • Activating cocktail to induce cytokine production
Kit Includes
  • UltraComp eBeads™ Plus Compensation Beads (01-3333-41)
  • CD16/CD32 Monoclonal Antibody (93), eBioscience (14-0161-81)
  • CellBlox™ Blocking Buffer (B001T06F01)
  • eBioscience Intracellular Fixation & Permeabilization Buffer Set (88-8824-00)
  • eBioscience™ Flow Cytometry Staining Buffer (00-4222-57)
  • eBioscience Cell Stimulation Cocktail plus protein transport inhibitors, 500X (00-4975-93)
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
For Use With (Application) Flow Cytometry