Learn More
STEMCELL Technologies EasySep™ Mouse NK Cell Isolation Kit
Description
- The EasySep™ Mouse NK Cell Isolation Kit targets non-NK cells by labeling unwanted cells with biotinylated antibodies and EasySep™ Streptavidin RapidSpheres™, and separates cells without columns using an EasySep™ magnet
- Desired cells are simply poured off into a new tube
- Isolated cells are immediately available for downstream applications such as flow cytometry, culture or cell-based assays
- This product replaces the EasySep™ Mouse NK Cell Enrichment Kit (Catalog #19755) for even faster cell isolations
Specifications
Specifications
| Cell Type | NK Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 cells |