Learn More
STEMCELL Technologies EasySep™ Mouse CD4+CD25+ Regulatory T Cell Isolation Kit II
Product Code. 17177871
Encompass
Change view
Click to view available options
Quantity:
1 x 109 Cells
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17177871 1 x 109 Cells 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17177871 Supplier STEMCELL Technologies Supplier No. 18783
Immunomagnetic isolation of mouse CD4+CD25+ regulatory T cells
  • The EasySep™ Mouse CD4+CD25+ Regulatory T Cell Isolation Kit II is designed to isolate CD4+CD25+ T cells from single-cell suspensions of splenocytes or other tissues by negative followed by positive selection
  • CD4+ T cells are first enriched by negative selection using the EasySep™ Mouse CD4+ T Cell Isolation Cocktail
  • CD25+ cells are then positively selected from the pre-enriched cells using EasySep™ Mouse CD25 Regulatory T Cell Positive Selection Cocktail
Specifications
Cell Type T Cells, T Cells, Regulatory
Quantity 1 x 109 Cells