Learn More
3M™ Easy Trap™ Easy Trap Duster Sheets
Product Code. 17108710
Change view
Click to view available options
Dimensions (L x W):
127 x 152 mm
Unit Size:
2 rolls
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
17108710 127 x 152 mm 2 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17108710 Supplier 3M™ Easy Trap™ Supplier No. 7100104251
  • ONLINE EXCLUSIVE PRICE
    193.00 EUR
    90.62 EUR / 2 rolls
    Save 102.38 EUR
    53% Off
Estimated Shipment: 30-12-2026
For dust, dirt, and sand removal
  • Traps and holds six times more dust, dirt, sand, and hair than traditional mops
  • Low drag reduces worker fatigue
  • Works in wet environments, and on most horizontal surfaces
  • Boosts productivity without compromising cleaning standards
  • Perforated rolls allow users to customise the sheet size, reducing waste
  • Helps to reduce the risk of spreading germs and bacteria
  • Contains a minimum of 60% recycled content
  • Includes 250 sheets per box
TRUSTED_SUSTAINABILITY
Specifications
Product Type Duster Sheets
Dimensions (L x W) 127 x 152 mm
For Use With (Application) Removal of Dust, Dirt, and Sand