Learn More
3M™ Easy Trap Duster Sheets
Product Code. 17287790
Change view
Click to view available options
Dimensions (L x W):
127 x 152 mm
203 x 152 mm
Unit Size:
1 roll
8 rolls
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
17287790 203 x 152 mm 1 roll
17297790 127 x 152 mm 8 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17287790 Supplier 3M™ Supplier No. 7100081546
  • ONLINE EXCLUSIVE PRICE
    230.00 EUR
    181.34 EUR / 1 rolls
    Save 48.66 EUR
    21% Off
View all promotions
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
Trap and hold eight times more dust, dirt, sand and hair than traditional mops and other disposable dusting systems. Two sided disposable sheets, works in wet environments and on most horizontal surfaces. Low drag reduces worker fatigue.
  • Duster system traps and holds six times more dust, dirt, sand and hair than traditional mops
  • Low drag reduces worker fatigue
  • Works in wet environments
  • Works on most horizontal surfaces
  • Two sided disposable sheets for use with your current dusting tools
TRUSTED_SUSTAINABILITY
Specifications
Product Type Duster Sheets
Dimensions (L x W) 203 x 152 mm
Quantity 60 Sheets/Roll, 8 Rolls/Cs.