missing translation for 'onlineSavingsMsg'
Learn More

(E)-[6]-Dehydroparadol, MedChemExpress

Product Code. p-200100711
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484117 50 mg 50mg
16467956 100 mg 100mg
16461956 500 mg 500mg
16440668 5 mg 5mg
16438228 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16484117 Supplier Medchem Express Supplier No. HY77293
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress (E)-[6]-Dehydroparadol, an oxidative metabolite of [6]-Shogaol (HY-14616), is a potent Nrf2 activator. (E)-[6]-Dehydroparadol can inhibit the growth and induce the apoptosis of human cancer cells.

(E)-[6]-Dehydroparadol, an oxidative metabolite of [6]-Shogaol (HY-14616), is a potent Nrf2 activator. (E)-[6]-Dehydroparadol can inhibit the growth and induce the apoptosis of human cancer cells.

Specifications

Chemical Name or Material (E)-[6]-Dehydroparadol
CAS 878006-06-5
Color Orange
Purity Grade Notes Research
Molecular Formula C17H24O3
Quantity 50 mg
Solubility Information DMSO : ≥ 140 mg/mL (506.57 mM)
SMILES CCCCCCCC(/C=C/C1=CC=C(O)C(OC)=C1)=O
Molecular Weight (g/mol) 276.37
Formula Weight 276.37
Percent Purity 95.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.