Learn More
(E)-4-Hydroxycrotonic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | (2E)-4-Hydroxy-2-butenoic Acid |
| CAS | 24587-49-3 |
| Molecular Formula | C4 H6 O3 |
| Quantity | 250 mg |
| Synonym | (E)-4-Hydroxycrotonic Acid, 2-Butenoic acid, 4-hydroxy-, (2E)-, 2-Butenoic acid, 4-hydroxy-, (E)-, Crotonic acid, 4-hydroxy-, (E)- (8CI), (2E)-4-Hydroxy-2-butenoic acid, (E)-4-Hydroxybut-2-enoic acid, (E)-4-Hydroxycrotonic acid, trans-4-Hydroxycrotonic acid, trans-γ-Hydroxycrotonic |
| InChI Formula | InChI=1S/C4H6O3/c5-3-1-2-4(6)7/h1-2,5H,3H2,(H,6,7)/b2-1+ |
| SMILES | OC\C=C\C(=O)O |
| IUPAC Name | (E)-4-hydroxybut-2-enoic acid |
| Molecular Weight (g/mol) | 102.089 |
| Formula Weight | 102.032 |