Learn More
(E)-3-(4-Amino-3,5-dimethylphenyl)acrylonitrile Hydrochloride, TRC
Product Code. 30309605
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30309605 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30309605 Supplier Toronto Research Chemicals Supplier No. TRCA6073402.5MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2E)-3-(4-Amino-3,5-dimethylphenyl)prop-2-enenitrile Hydrochloride
CAS 661489-23-2
Molecular Formula C11 H12 N2 . Cl H
Quantity 2.5 mg
Synonym 2-Propenenitrile, 3-(4-amino-3,5-dimethylphenyl)-, hydrochloride (1:1), (2E)-, 2-Propenenitrile, 3-(4-amino-3,5-dimethylphenyl)-, monohydrochloride, (2E)- (9CI), (E)-3-(4-Amino-3,5-dimethylphenyl)-2-propenenitrile hydrochloride
InChI Formula InChI=1S/C11H12N2.ClH/c1-8-6-10(4-3-5-12)7-9(2)11(8)13;/h3-4,6-7H,13H2,1-2H3;1H/b4-3+;
SMILES Cl.Cc1cc(\C=C\C#N)cc(C)c1N
IUPAC Name (E)-3-(4-amino-3,5-dimethylphenyl)prop-2-enenitrile;hydrochloride
Molecular Weight (g/mol) 208.69
Formula Weight 208.0767