missing translation for 'onlineSavingsMsg'
Learn More

E 2012, MedChemExpress

Product Code. 16472808
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
This item is not returnable. View return policy

Product Code. 16472808

Brand: Medchem Express HY10016

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress E 2012 is a potent gamma (γ) secretase modulator without affecting Notch processing. E 2012 inhibits 3β-hydroxysterol δ24-reductase (DHCR24) at the final step in the cholesterol biosynthesis. E 2012 aims at Alzheimer's disease by reduction of amyloid β-42, and induces cataract following repeated doses in the rat.

E 2012 is a potent gamma (γ) secretase modulator without affecting Notch processing. E 2012 inhibits 3β-hydroxysterol δ24-reductase (DHCR24) at the final step in the cholesterol biosynthesis. E 2012 aims at Alzheimer's disease by reduction of amyloid β-42, and induces cataract following repeated doses in the rat.

Specifications

Chemical Name or Material E 2012
CAS 870843-42-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C25H26FN3O2
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (119.19 mM)
SMILES O=C(N([C@H](C1=CC=C(C=C1)F)C)CCC/2)C2=C\C3=CC=C(N4C=NC(C)=C4)C(OC)=C3
Molecular Weight (g/mol) 419.49
Formula Weight 419.49
Percent Purity 97.39%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.