Learn More
(E)-1-Methoxy-4-(prop-1-en-1-yl)benzene, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 4180-23-8 |
| Molecular Formula | C10H12O |
| MDL Number | MFCD00009284 |
| Quantity | 1 kg |
| SMILES | C/C=C/C1=CC=C(OC)C=C1 |
| Molecular Weight (g/mol) | 148.20 |
| Percent Purity | 97% |
| Chemical Name or Material | (E)-1-Methoxy-4-(prop-1-en-1-yl)benzene |