Learn More
ABEBA™ Dynamic White Occupational Clogs Ob ESD
Product Code. 17151365
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17151365 39 1 pair
17111365 35 1 pair
17121365 36 1 pair
17111375 45 1 pair
17121375 46 1 pair
17131375 47 1 pair
17141375 48 1 pair
17131365 37 1 pair
17141365 38 1 pair
17161365 40 1 pair
17171365 41 1 pair
17181365 42 1 pair
17191365 43 1 pair
17101375 44 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 17151365 Supplier ABEBA™ Supplier No. 37329/39
Estimated Shipment: 21-12-2026
White dynamic occupational clogs with non-foldable heel strap, replaceable active comfort insole, slip resistant TPU outsole
  • Breathable, lightweight, comfortable occupational clogs
  • Comply to EN ISO 20347: 2012, OB SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant TPU outsole
  • Good cushioning
  • Energy absorption in heel area
  • Water repellent white knitted textile
  • Non foldable heel strap, adjustable
  • Removable active comfort insole
Specifications
Color White
Material Knitted Fabric
Size 39
Certifications/Compliance (Europe) ESD dissipative to EN 61340 Climate class 1, Comply to EN ISO 20347: 2012 OB SRC
Gender Unisex
Ankle Style Clogs
Waterproof Water Repellent
Product Type OB SRC ESD