Learn More
Fisherbrand™ Dycem™ Polymeric Compound Non-slip Mats
Product Code. 12846445 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Dimensions (L x W):
200 x 200 mm
320 x 260 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
12846445 200 x 200 mm Each
12856445 320 x 260 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12846445
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
Estimated Shipment: 10-03-2027
Add to basket
Add to basket
Provides a secure surface that anchors items such as cups, plates, tools, and a variety of other objects.
  • Can be washed with soap and water
  • Latex-free
Specifications
Color Blue
Dimensions (L x W) 200 x 200 mm
Material Dychem Polymeric Compound