Learn More
DWK Life Sciences DURAN™ TILT Bottle Light Shield, White, Silicone Rubber
Shop All DWK Life Sciences Products
Click to view available options
:
White silicone
Description
The DURAN™ TILT Light Shield is a white silicone sleeve that covers 94% of the bottle surface. The sleeve has a number of protective functions: it blocks damaging ultraviolet light (UV), protects the glass surface from damage, and facilitates safer handling. DURAN™ TILT Light Shield includes four GL 56 Bottle Tags (Orange, Yellow, Blue, Purple) made from silicone.
- The white DURAN™ TILT Light Shield is a silicone sleeve that covers 94 % of the bottle surface and blocks damaging ultraviolet light
- Protects the glass surface from damage
- Facilitates safer handling
- Silicone provides a much better grip than glass especially when the bottle has been sprayed with 70 % alcohol for surface disinfection
Specifications
Specifications
| Color | White |
| Material | Silicone |
| For Use With (Application) | Working with cell culture media under sterile conditions in biosafety cabinets and clean hoods; Storage of light-sensitive cell culture media |
| No. per Pack | 4/Pk. |
| Autoclavable | Autoclavable |