Learn More
DWK Life Sciences DURAN™ Multi-purpose Cylinder, with round base, without graduation
Shop All DWK Life Sciences ProductsDescription
The multitude of different work steps in the laboratory environment demands a multitude of universally applicable glassware. These glass devices usually have a very simple shape and are characterized by their versatility.
- Rough ground rim
- With round base
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| Diameter (Metric) Outside | 50 mm |
| For Use With (Application) | Diluting solutions; Mixing several components |
| Capacity (Metric) | 770 mL |
| Autoclavable | Autoclavable |
| Quantity | 10/Pk. |