Learn More
Lenz Laborglasinstrumente™ Duran™ Medium-neck Round Bottom Flasks
Product Code. 10211551
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Diameter (Metric) Outer:
31 mm
45 mm
51 mm
64 mm
85 mm
105 mm
131 mm
166 mm
Standard Taper Joint:
NS 14/23
NS 29/32
Unit Size:
Each
Pack of 10
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Outer Standard Taper Joint Unit Size
10211551 250 mL 85 mm NS 29/32 Pack of 10
10483861 1000 mL 131 mm NS 29/32 Pack of 10
15360895 10 mL 31 mm NS 14/23 Pack of 10
10559421 100 mL 64 mm NS 29/32 Each
11737425 100 mL 64 mm NS 29/32 Pack of 10
10589611 2000 mL 166 mm NS 29/32 Each
15370885 25 mL 45 mm NS 14/23 Pack of 10
11915956 500 mL 105 mm NS 29/32 Pack of 10
10220831 500 mL 105 mm NS 29/32 Each
10710992 50 mL 51 mm NS 14/23 Pack of 10
10508851 50 mL 51 mm NS 29/32 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 10211551 Supplier Lenz Laborglasinstrumente™ Supplier No. 03002949
  • ONLINE EXCLUSIVE PRICE
    201.38 EUR
    135.73 EUR / Pack of 10
    Save 65.65 EUR
    32% Off
Estimated Shipment: 14-12-2026
Manufactured from Duran™ glass. Lenz Laborglasinstrumente™ Duran™ Medium-neck Round Bottom Flasks include a conical ground joint.
  • With inscription label
  • Acc. to DIN EN ISO 4797
  • Corrosion-free and resistant to thermal shock
  • With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Certifications/Compliance DIN EN ISO 4797
Capacity (Metric) 250 mL
Material Borosilicate Glass, Duran Glass
Neck Style Medium
Standard Taper Joint NS 29/32
Bottom Shape Round
Diameter (Metric) Outer 85 mm