Learn More
WTW™ DURAN™ Measuring Bottle
Product Code. 10400103
Change view
Click to view available options
Volume (Metric) Sample:
500mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sample Unit Size
10400103 500mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10400103 Supplier WTW™ Supplier No. 209107
  • ONLINE EXCLUSIVE PRICE
    242.00 EUR
    227.42 EUR / Pack of 1
    Save 14.58 EUR
    6% Off
Estimated Shipment: 15-12-2026
WTW offers a wide variety of measuring bottles and measuring vessels with adapters to measure BSB, soil respiration, degradeability as per OECD 301F, fermentation tests, biogas and much more. They have all be tested in their respective applications and therefore support accurate measurements.
TRUSTED_SUSTAINABILITY
Specifications
Type Measuring Bottle Duran
Includes 2 Nozzles with 2 x VS25 Screwing Sets for Nozzle, 1 x GK 600 Rubber Sleeve, 2 x SEP25 Septum for Nozzles, 2 x STH25 Support Sleeves for Nozzle
Volume (Metric) Sample 500mL