Learn More
DWK Life Sciences DURAN™ GLS 80™ Screw Cap with Four Ports
Product Code. 15710239 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
GLS 80
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) Unit Size
15710239 GLS 80 Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15710239 Supplier DWK Life Sciences Supplier No. 293109103
  • ONLINE EXCLUSIVE PRICE
    216.00 EUR
    176.59 EUR / Pack of 2
    Save 39.41 EUR
    18% Off
Estimated Shipment: 22-12-2026
The DURAN™ GLS 80™ Screw Cap with Four Ports provides a flexible, modular system for safe liquid and gas transfer within closed, sterile environments. Compatible with five tubing diameters (3.2 to 12.0 mm) and fully compatible with all GLS 80 threads.
  • Flexible modular GLS 80 connection system for safe transfer of liquid or gas media within a closed and sterile environment
  • Four ports with GL 18 thread accept five interchangeable insert diameters: 3.2 mm, 6.0 mm, 8.0 mm, 10.0 mm and 12.0 mm
  • Tubes are fed through ports and secured with inserts and screw connections
  • Sterile pressure equalisation is possible by using the syringe membrane filter (pressure equalisation kit available separately)
  • Unused ports can be sealed with a closed-top GL 18 screw cap (blanking cap)
  • Fully compatible with all GLS 80 threads, offering high versatility across the DURAN™ bottle range
  • Constructed from PP and PTFE; reduces evaporation during media transfer
Specifications
Color Blue
Description DURAN GLS 80 screw cap with four ports (GL 18 thread) Blue and grey Polypropylene with white EPDM O-ring
For Use With (Application) Fluid transfer; Supply of liquid into bioreactors; Supply of reagents to instruments; Photo-bioreactor
Material Polypropylene, PTFE, EPDM
Quantity 2/Pk.
Sterility Non-sterile
Thread Size (Metric) GLS 80
Neck Finish 93 mm
Diameter (Metric) 87 mm
Autoclavable Autoclavable
Disposable Reusable
Product Type Bottle Cap