Learn More
DWK Life Sciences DURAN™ GLS 80™ Pouring Ring, Blue PP
Product Code. 10611724 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
GLS 80
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) Unit Size
10611724 GLS 80 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10611724 Supplier DWK Life Sciences Supplier No. 1160166
  • ONLINE EXCLUSIVE PRICE
    27.70 EUR
    19.87 EUR / Pack of 10
    Save 7.83 EUR
    28% Off
Estimated Shipment: 30-12-2026
Blue PP GLS 80™ wide-mouth screw cap (plug seal) and drip free, PP pouring ring for GLS 80™ bottles
Permits opening and closing of the DURAN™ GLS 80™ bottle with only a three-quarter turn. A matching PP pouring ring is also available, permitting clean, drip-free use.
  • Triple thread permits user friendly opening and closing with only a three-quarter turn
  • Cap offer a linerless, integral lip seal
  • Pouring ring for drip-free pouring and clean, safe working
Specifications
Description Pouring ring
For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping
Material Polypropylene
Quantity 10/Pk.
Thread Size (Metric) GLS 80
Neck Finish 16 mm
Diameter (Metric) 87 mm