Learn More
DWK Life Sciences DURAN™ Glass Stopcock for Aspirator Bottles, Standard Taper Ground Cone
Product Code. 10686161 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
5000 to 20,000 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10686161 5000 to 20,000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10686161 Supplier DWK Life Sciences Supplier No. 241480401
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Stopcock with glass key and a ground cone for Aspirator bottles
Aspirator stopcock with glass key and a ground cone to fit the socket side-arm of DURAN™ aspirator bottles.
  • All glass construction
  • Tight closure due to standard ground joint
Specifications
Capacity (Metric) 5000 to 20,000 mL
For Use With (Application) Controlled dispensing of liquids and reagents
Material Borosilicate Glass (3.3)
No. per Pack 1/Pk.