Learn More
Lenz Laborglasinstrumente™ Double Walled Coil Condensers with GL Threads
Product Code. 10121602
Change view
Click to view available options
Length (Metric) Jacket:
250 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Jacket Unit Size
10121602 250 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10121602 Supplier Lenz Laborglasinstrumente™ Supplier No. 05250314
  • ONLINE EXCLUSIVE PRICE
    158.00 EUR
    105.33 EUR / Pack of 1
    Save 52.67 EUR
    33% Off
Estimated Shipment: 22-12-2026
Facilitates the condensing of vapors in the laboratory. Lenz Laborglasinstrumente™ Double Walled Coil Condensers with GL Threads are manufactured from durable DURAN™ tubing.
  • Acc. to DIN 12593
  • With NS socket and cone
  • With cooling coil and jacket
  • Corrosion-free and resistant to thermal shock
  • Water connection: GL 14 threads (with two threaded plastic olives)
Specifications
Length (Metric) Jacket 250 mm
Quantity 1 Each
Material Duran Glass
Type Intensive Condenser