Learn More
Lenz Laborglasinstrumente™ Duran Glass Columns acc. to Vigreux
Product Code. 10181421
Change view
Click to view available options
Standard Taper Joint:
NS 14/23
NS 24/29
NS 29/32
Length (Metric):
200mm
300mm
600mm
Includes:
NS socket and NS cone
With removable glass jacket
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Length (Metric) Includes Unit Size
10181421 NS 14/23 200mm NS socket and NS cone Each
10346261 NS 14/23 300mm With removable glass jacket Each
10396161 NS 24/29 300mm NS socket and NS cone Each
10747141 NS 14/23 300mm NS socket and NS cone Each
10234101 NS 29/32 300mm NS socket and NS cone Each
10355641 NS 29/32 300mm With removable glass jacket Each
10632282 NS 29/32 600mm With removable glass jacket Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10181421 Supplier Lenz Laborglasinstrumente™ Supplier No. 05130201
  • ONLINE EXCLUSIVE PRICE
    65.00 EUR
    59.81 EUR / Pack of 1
    Save 5.19 EUR
    7% Off
Estimated Shipment: 29-10-2026
Specifications
Includes NS socket and NS cone
Length (Metric) 200mm
Type Columns acc. to Vigreux
Material Duran Glass
Quantity 1Each
Standard Taper Joint NS 14/23