Learn More
Lenz Laborglasinstrumente™ Duran™ Gas Sampling Tube
Product Code. 10737181
Change view
Click to view available options
Capacity (Metric):
150 mL
350 mL
500 mL
1000 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10737181 1000 mL Each
10221851 150 mL Each
10057970 500 mL Each
10796981 350 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10737181 Supplier Lenz Laborglasinstrumente™ Supplier No. 05680170
  • ONLINE EXCLUSIVE PRICE
    187.00 EUR
    123.60 EUR / Pack of 1
    Save 63.40 EUR
    33% Off
Estimated Shipment: 17-09-2026
Specifications
Capacity (Metric) 1000 mL
Material Duran Glass
Type Gas Sampling Tube
Includes Nozzle GL14 Thread and septa
Certifications/Compliance DIN 12473-1
Quantity 1 Pack