Learn More
Lenz Laborglasinstrumente™ Duran™ Extractors with Allihn Condenser
Product Code. 10624012
Change view
Click to view available options
Capacity (Metric):
100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10624012 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10624012 Supplier Lenz Laborglasinstrumente™ Supplier No. 05365037
  • ONLINE EXCLUSIVE PRICE
    286.00 EUR
    234.62 EUR / Pack of 1
    Save 51.38 EUR
    17% Off
Estimated Shipment: 14-12-2026
Specifications
For Use With (Equipment) Allihn Condenser
Product Type Extractor
Capacity (Metric) 100 mL
Material Duran Glass
Includes Allihn condenser
For Use With (Application) For the extraction from solids with solvents
Thread Size GL 14