Learn More
Lenz Laborglasinstrumente™ Duran™ Extractors
Product Code. 10587321
Change view
Click to view available options
Capacity (Metric):
100 mL
200 mL
250 mL
1000 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10587321 1000 mL Each
10615051 100 mL Each
10652361 200 mL Each
11806194 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10587321 Supplier Lenz Laborglasinstrumente™ Supplier No. 05360070
  • ONLINE EXCLUSIVE PRICE
    555.00 EUR
    441.34 EUR / Pack of 1
    Save 113.66 EUR
    20% Off
Estimated Shipment: 05-10-2026
Specifications
For Use With (Equipment) Dimroth Condenser
Product Type Extractor
Capacity (Metric) 1000 mL
Material Duran Glass
Includes Dimroth condenser
For Use With (Application) For the extraction from solids with solvents
Thread Size GL 14