Learn More
Lenz Laborglasinstrumente™ Duran™ Extractor Heads with NS 2 Way Stopcock
Product Code. 10612092
Change view
Click to view available options
Capacity (Metric):
30 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10612092 30 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10612092 Supplier Lenz Laborglasinstrumente™ Supplier No. 05405125
  • ONLINE EXCLUSIVE PRICE
    171.00 EUR
    130.41 EUR / Pack of 1
    Save 40.59 EUR
    23% Off
Estimated Shipment: 14-12-2026