Learn More
DWK Life Sciences DURAN™ Evaporating Flask, with standard ground joint, pear shape
Product Code. 15251859 Shop All DWK Life Sciences Products
Change view
Click to view available options
Volume (Metric):
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
15251859 250 mL Pack of 10
15241859 100 mL Pack of 10
15261859 500 mL Pack of 10
15271859 1000 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 15251859 Leverandør DWK Life Sciences Leverandørnr. 241203703
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy
Evaporating flasks are particularly suitable for evaporating liquids or solvents.
Evaporating flasks are a variant of round flasks and have a pear-shaped shape. The standard ground joint allows a precise connection to other ground joint components, e.g. a rotary evaporator. Due to their shape, evaporating flasks allow a large uniform liquid surface, making them particularly suitable for the evaporation of solvents.
  • Tight closure due to standard ground joint
  • Fired-on, highly durable white print
  • Pear shape
TRUSTED_SUSTAINABILITY
Tekniske data
Description Autoclavable
Material Borosilicate Glass (3.3)
For Use With (Application) Evaporation of liquids; Heating; Suitable for heating baths and mantles; Mixing several components; Rotary evaporator flask, suitable for all standard rotary evaporators; Concentration of solutions
Quantity 10/Pk.
Volume (Metric) 250 mL
Diameter (Metric) 81 mm
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.